Specifications
| Quantity | 1 g |
| Melting Point | 191.0°C |
| Formula Weight | 356.33 |
| InChI Key | DUAGQYUORDTXOR-GPQRQXLASA-N |
| SMILES | OC[C@H]1O[C@@H](O[C@@H]2OC=C3C(=O)OCC=C3[C@H]2C=C)[C@H](O)[C@@H](O)[C@@H]1O |
| IUPAC Name | (5R,6S)-5-ethenyl-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-1H,3H,5H,6H-pyrano[3,4-c]pyran-1-one |
| Molecular Weight (g/mol) | 356.33 |
| Chemical Name or Material | Gentiopicroside |