missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gentiopicroside, MedChemExpress
Description
Gentiopicroside, a naturally occurring iridoid glycoside, inhibits P450 activity, with an IC50 and a Ki of 61 μM and 22.8 μM for CYP2A6
Specifications
Specifications
| Chemical Name or Material | Gentiopicroside |
| CAS | 20831-76-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H20O9 |
| Quantity | 25 mg |
| Synonym | Gentiopicrin |
| Solubility Information | DMSO : ≥ 100 mg/mL (280.65 mM) ∣H2O : 100 mg/mL (280.65 mM; Need ultrasonic) |
| SMILES | C=C[C@@H]([C@@H]1O[C@]([C@@H]([C@@H](O)[C@@H]2O)O)([H])O[C@@H]2CO)C(C3=CO1)=CCOC3=O |
| Molecular Weight (g/mol) | 356.32 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?