Learn More
Thermo Scientific™ Gentamicin Sulfate, Ultrapure
Product Code. 15815148 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
5 g
25 g
Form:
Powder
Unit Size:
1g
25g
5g
Product Code. 15815148
Brand: Thermo Scientific™ J16051.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Inhibits normal protein synthesis
  • Bactericidal
  • Stable
  • Autoclavable
  • Effective when combined with antibiotic that interferes with cell wall synthesis
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Requires no refrigeration
Antibiotic Spectrum Bactericidal
Concentration ≥590 μg/mg (Dry basis)
Form Powder
Product Type Antibiotic
Format Plastic bottle
pH 3.5 to 5.5
Purity or Quality Grade Ultrapure
Quantity 25 g