missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Genipin 1-β-D-gentiobioside, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Description
Genipin 1-β-D-gentiobioside (Genipin 1-gentiobioside) is one of the most abundant and bioactive iridoid glycosides in Gardenia jasminoides Ellis, which possesses hepatoprotective, anti-inflammatory, antioxidant, and antithrombotic activities.
Specifications
Specifications
| Chemical Name or Material | Genipin 1-β-D-gentiobioside |
| CAS | 29307-60-6 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C23H34O15 |
| Quantity | 25 mg |
| Synonym | Genipin 1-gentiobioside Genipin 1-β-gentiobioside Genipin gentiobioside |
| Solubility Information | Ethanol : 125 mg/mL (227.06 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (181.65 mM; Need ultrasonic) |
| SMILES | O[C@H]([C@@H](O)[C@@H]1O)[C@](O[C@@H]1CO[C@@H]([C@@H]([C@@H](O)[C@@H]2O)O)O[C@@H]2CO)([H])O[C@H]3[C@@](C(CO)=CC4)([H])[C@@]4([H])C(C(OC)=O)=CO3 |
| Molecular Weight (g/mol) | 550.51 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction