missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneScan™ 500 TAMRA™ dye Size Standard

Product Code. 16375242 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
800 reactions
Unit Size:
800 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16375242 800 reactions 800 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16375242 Supplier Applied Biosystems™ Supplier No. 401733

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

This kit contains 16 TAMRA dye labeled single stranded DNA oligos for size standards.

GeneScan™ 500 TAMRA™ Size Standard is an internal lane size standard developed for use with the Applied Biosystems fluorescence- based DNA electrophoresis systems.

  • The use of an internal lane size standard during electrophoresis enables automated data analysis and precise DNA fragment size comparisons between electrophoresis runs

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Product Line GeneScan, TAMRA
Content And Storage This kit contains 16 TAMRA dye labeled single stranded DNA oligos for size standards. Store kit at 2-8°C. (Do not freeze)
For Use With (Equipment) 310 Genetic Analyzer, 3130xl Genetic Analyzer, 3730xl DNA Analyzer, SeqStudio Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
Label or Dye TAMRA™
Quantity 800 reactions
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.