missing translation for 'onlineSavingsMsg'
Learn More

GDF-9 Antibody (GDF9/4261), DyLight 488, Novus Biologicals™

Product Code. 18589002 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18589002 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18589002 Supplier Novus Biologicals Supplier No. NBP308362G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GDF-9 Monoclonal antibody specifically detects GDF-9 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GDF-9
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GDF9/4261
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9
Host Species Mouse
Immunogen Tuberculin coupled peptide with sequence VPAKYSPLSVLTIEPDGSIAYKEYEDMIATKC that recognizes an epitope with the EPDG sequence near the C-terminal region of human GDF-9. (Uniprot: O60383)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 2661
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.