missing translation for 'onlineSavingsMsg'
Learn More

Gcase activator 1, MedChemExpress

Product Code. 16453938
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453938

Brand: Medchem Express HY104038

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gcase activator 1 is an activator of glucocerebrosidase (Gcase) extracted from patent WO 2017192841 A1.

Gcase activator 1 is an activator of glucocerebrosidase (Gcase) extracted from patent WO 2017192841 A1.

Specifications

Chemical Name or Material Gcase activator 1
Physical Form Solid
CAS 1919820-28-2
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H30N4O2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 50 mg/mL (139.48 mM; Need ultrasonic)
SMILES O=C(C1=C2N=C(C)C=C(C)N2N=C1)N[C@@H]3CC[C@@H](OCCCCC)CC3
Molecular Weight (g/mol) 358.48
Formula Weight 358.48
Percent Purity 98.77%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.