Learn More
Trajan Scientific and Medical™ Replacement Mini Union Ferrules
Product Code. 12736348
Change view
Click to view available options
For Use With (Equipment):
0.1 to 0.32 mm I. D Columns
0.45 to 0.53 mm ID Columns
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12736348 0.45 to 0.53 mm ID Columns Pack of 10
12746348 0.1 to 0.32 mm I. D Columns Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12736348 Supplier Trajan Scientific and Medical™ Supplier No. 072629
Please to purchase this item. Need a web account? Register with us today!
Leak-free system connections for improved chromatography for non-MS systems
  • Easy to use
  • Forms a reliable seal
  • Reusable
  • Soft material, forms a soft grip to the capillary column
  • Low emissions
Specifications
Type Ferrule
For Use With (Equipment) 0.45 to 0.53 mm ID Columns
Material Graphite
Quantity 10 Pack