Learn More
Gboxin, MedChemExpress
Product Code. 16494508
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494508 10 mg 10mg
16480817 50 mg 50mg
16437227 5 mg 5mg
16405376 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16494508 Supplier Medchem Express Supplier No. HY111651
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Gboxin is an oxidative phosphorylation (OXPHOS) inhibitor that targets glioblastoma. Gboxin inhibits the activity of F0F1 ATP synthase. Antitumour activity.
Gboxin is an oxidative phosphorylation (OXPHOS) inhibitor that targets glioblastoma. Gboxin inhibits the activity of F0F1 ATP synthase. Antitumour activity.
Specifications
Chemical Name or Material Gboxin
CAS 2101315-36-8
Color White
Purity Grade Notes Research
Molecular Formula C22H33ClN2O2
Quantity 10 mg
Solubility Information DMSO : 32.5 mg/mL (82.71 mM; Need ultrasonic) ∣H2O : 10 mg/mL (25.45 mM; Need ultrasonic)
SMILES O=C(O[C@H]1[C@H](C(C)C)CC[C@@H](C)C1)CN2C(CC)=[N+](C)C3=CC=CC=C23.[Cl-]
Molecular Weight (g/mol) 392.96
Formula Weight 392.96
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease