missing translation for 'onlineSavingsMsg'
Learn More

GBL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

216.00 € - 326.00 €

Specifications

Antigen GBL
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1-5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18609111
View Documents
Novus Biologicals
NBP2-98484-100ul
100 μg
326.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18618914
View Documents
Novus Biologicals
NBP2-98484-50ul
50 μg
216.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GBL Polyclonal antibody specifically detects GBL in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications

Specifications

GBL
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Hypoxia, mTOR Pathway
PBS (pH 7.0)
64223
IgG
Antigen and protein A Affinity-purified
Western Blot 1:500-1:2000, Immunoprecipitation 1-5 μL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200
Polyclonal
Purified
RUO
Human
G protein beta subunit-like, Gable, GbetaL, GBLPop3, Lst8, LST8MGC111011, Mammalian lethal with SEC13 protein 8, mLST8, MTOR associated protein, LST8 homolog (S. cerevisiae), POP3, Protein GbetaL, target of rapamycin complex subunit LST8, TORC subunit LST8, WAT1
Produced in rabbits immunized with E. coli-derived Human GBL fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.