missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBE1 Antibody (OTI1D11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-02245
This item is not returnable.
View return policy
Description
GBE1 Monoclonal specifically detects GBE1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| GBE1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:50-100, Immunoprecipitation 2ug/500ul, Immunohistochemistry-Paraffin 1:50 | |
| GBE1 | |
| Full length human recombinant protein of human GBE1 (NP_000149) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 2632 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| OTI1D11 | |
| Unconjugated | |
| 1,4-alpha-glucan-branching enzyme, amylo-(1,4 to 1,6) transglucosidase, amylo-(1,4 to 1,6) transglycosylase, Brancher enzyme, EC 2.4.1.18, GBE, glucan (1,4-alpha), branching enzyme 1, glycogen branching enzyme, Glycogen-branching enzyme | |
| Mouse | |
| 80.3 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction