Description
- Material: Borosilicate glass 3.3
- Type: Gay-Lussac
- DIN ISO 3507
- Calibrated to contain (TC, In)
- Includes individual certificate
- Stopper NS 10/19 with capillary, top of stopper ground and polished
Specifications
Specifications
| Calibration | To Contain |
| Capacity (Metric) | 10 mL |
| Material | Borosilicate Glass (3.3) |
| Shape | Gay-Lussac |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Graduated | Yes |
| Mouth Style | Narrow |
| Sterility | Non-sterile |
| Bottom Shape | Flat |