missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA-3 Mouse anti-Human, Clone: GATA3/2446, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07704-20UG
This item is not returnable.
View return policy
Description
GATA-3 Monoclonal specifically detects GATA-3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications
| GATA-3 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA | |
| 131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3 | |
| Recombinant fragment of human GATA-3 protein (around aa 357-436) (exact sequence is proprietary) | |
| 20 μg | |
| Cancer, Innate Immunity | |
| 2625 | |
| Store at 4°C. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, ELISA | |
| GATA3/2446 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction