missing translation for 'onlineSavingsMsg'
Learn More

GATA-3 Antibody (GATA3/4550R) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30173009
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30173009 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30173009 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP314267JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GATA-3 Monoclonal antibody specifically detects GATA-3 in Human,Rat samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GATA-3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GATA3/4550R
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias 131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3
Host Species Rabbit
Immunogen Recombinant full-length human GATA-3 protein (Uniprot: P23771)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 2625
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.