Learn More
Gastrodenol, MedChemExpress
Product Code. 16491068
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 16491068
Brand: Medchem Express HYB0796
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Gastrodenol(Bismuth tripotassium dicitrate; De-Noltab)is a mineral that is used in treating ulcers and upset stomach.
Gastrodenol(Bismuth tripotassium dicitrate
Specifications
Chemical Name or Material Gastrodenol
Physical Form Solid
CAS 57644-54-9
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C12H10BiK3O14
Synonym Bismuth tripotassium dicitrate Bismuth subcitrate
Solubility Information H2O : 100 mg/mL (141.95 mM; Need ultrasonic)
SMILES O=C(CC(C([O-])=O)(O)CC([O-])=O)[O-].O=C(CC(C([O-])=O)(O)CC([O-])=O)[O-].[K+].[K+].[K+].[Bi+3]
Molecular Weight (g/mol) 704.47
Formula Weight 704.47
Percent Purity 95.0%
Grade Research