Learn More
WLD Tec X2 Gas cartridge CV470, 450gr butane with valve .
Brand:  WLD Tec 6.000.185
X2 Gas cartridge CV470, 450gr butane with valve .
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10149311
  • ONLINE EXCLUSIVE PRICE
    65.10 €
    37.19 € / Pack of 2
    Save 27.91 €
    42% Off
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Camping Gaz Kartuschen
Signal Word
  • Danger
Hazard Category
  • Flammable gas.
Hazard Statement
  • H220-Extremely flammable gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P102-Keep out of reach of children.
  • P381-Eliminate all ignition sources if safe to do so.
  • P403-Store in a well-ventilated place.
  • P377-Leaking gas fire: Do not extinguish, unless leak can be stopped safely.