missing translation for 'onlineSavingsMsg'
Learn More

GAS8 Monoclonal Antibody (OTI10G2), TrueMAB™, OriGene

Product Code. 17540094
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17540094 30 μL 30µL
17520094 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17540094 Supplier Origene Technologies Supplier No. TA808629S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GAS8 Monoclonal antibody specifically detects GAS8 in Human samples. It is validated for Western Blot

Specifications

Antigen GAS8
Applications Western Blot
Classification Monoclonal
Clone OTI10G2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Cnot2
Gene Accession No. Q9NZN8
Gene Alias beta growth arrest specific protein 11; CILD33; DRC4; Dynein regulatory complex subunit 4; Gas11; GAS-11; Gas8; GAS-8; growth arrest specific 11; growth arrest specific 8; growth arrest specific protein 11; growth arrest-specific protein 11; Growth arrest-specific protein 8; MGC138326
Host Species Mouse
Immunogen Full length human recombinant protein of human CNOT2 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4848
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.