Learn More
Garenoxacin Mesylate hydrate, MedChemExpress
Product Code. 16469837
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16469837
Brand: Medchem Express HY17460A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Garenoxacin Mesylate hydrate (BMS284756 Mesylate hydrate) is a novel oral des-fluoro(6) quinolone with potent antimicrobial activity, against common respiratory pathogens, including resistant strains.
Garenoxacin Mesylate hydrate (BMS284756 Mesylate hydrate) is a novel oral des-fluoro(6) quinolone with potent antimicrobial activity, against common respiratory pathogens, including resistant strains.
Specifications
Chemical Name or Material Garenoxacin Mesylate hydrate
Physical Form Solid
CAS 223652-90-2
Color Ash Gray
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C24H26F2N2O8S
For Use With (Application) COVID-19-immunoregulation
Synonym BMS284756 Mesylate hydrate
Solubility Information DMSO : 100 mg/mL (185.00 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (23.13 mM; Need ultrasonic)
SMILES O=C(C1=CN(C2CC2)C3=C(C=CC(C4=CC5=C([C@@H](C)NC5)C=C4)=C3OC(F)F)C1=O)O.CS(=O)(O)=O.O
Molecular Weight (g/mol) 540.53
Formula Weight 540.53
Percent Purity 98.2%
Grade Research