missing translation for 'onlineSavingsMsg'
Learn More

Garenoxacin Mesylate hydrate, MedChemExpress

Product Code. 16437358
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16437358

Brand: Medchem Express HY17460A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Garenoxacin Mesylate hydrate (BMS284756 Mesylate hydrate) is a novel oral des-fluoro(6) quinolone with potent antimicrobial activity, against common respiratory pathogens, including resistant strains.

Garenoxacin Mesylate hydrate (BMS284756 Mesylate hydrate) is a novel oral des-fluoro(6) quinolone with potent antimicrobial activity, against common respiratory pathogens, including resistant strains.

Specifications

Chemical Name or Material Garenoxacin Mesylate hydrate
Physical Form Solid
CAS 223652-90-2
Color Ash Gray
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C24H26F2N2O8S
For Use With (Application) COVID-19-immunoregulation
Synonym BMS284756 Mesylate hydrate
Solubility Information DMSO : 100 mg/mL (185.00 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (23.13 mM; Need ultrasonic)
SMILES O=C(C1=CN(C2CC2)C3=C(C=CC(C4=CC5=C([C@@H](C)NC5)C=C4)=C3OC(F)F)C1=O)O.CS(=O)(O)=O.O
Molecular Weight (g/mol) 540.53
Formula Weight 540.53
Percent Purity 98.2%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.