Learn More
Ansell GAMMEX™ Polyisoprene Plus Glove-in-Glove System Surgical Gloves
Product Code. p-7250200 Shop All Ansell Products
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
Unit Size:
50 pairs
Product Code. 17843782
Brand: Ansell 340082060
Only null left
Add to basket
Add to basket
Synthetic, pre-donned double gloving system of two polyisoprene gloves in one, making it easy to double glove in one don for high risk surgical procedures.
  • ASTM D3577, ASTM D7160, ASTM F739 certified
  • High grip under dry and damp conditions
  • Softer and 'stretchier' polyisoprene glove featuring superior comfort compared to Ansell's latex gloves
  • Free from chemical accelerators known to cause chemical Type IV allergies and sensitivities
  • Outer and inner gloves are pre-donned and aligned at the finger tips for easy double gloving in one don, saving on half the donning time
  • 50% less inner packaging materials to help reduce the impact on the environment
  • Outer glove comes off easily for easy intra-operative changes
Specifications
Certifications/Compliance ASTM D3577, ASTM D7160, ASTM F739, ISO 10282
Certifications/Compliance (Europe) EN 16523-1, EN 374-2 and -4, EN 455 1-4, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-5:2016, EN 420:2003 + A1:2009, JIS T9107, AS/NZS 4179
Accelerator Free No
Applications Healthcare
AQL 0.65
Color White, Green
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 16.93 ±1.97 mil
Finger Thickness (Metric) 0.43 ±0.05 mm
Hand Specific
Laboratory/Cleanroom Use No
Latex Free Yes
Material Polyisoprene
Palm Thickness (English) 15.55 ±1.97 mil
Palm Thickness (Metric) 0.4 ±0.05 mm
Powdered Powder Free
Size 6
Sterility Sterile
Thickness (English) 15.55 ±1.97 mil
Thickness (Metric) 0.4 ±0.05 mm
Model GAMMEX-340082
Length (English) 12 in. (Outer), 11.9 in. (Inner)
Length (Metric) 305 mm (Outer), 303 mm (Inner)