Learn More
Gamma-methyl-L-leucine, TRC
Product Code. 30393814
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393814 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30393814 Supplier Toronto Research Chemicals Supplier No. TRCG195005100MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material gamma-methyl-L-leucine
CAS 57224-50-7
Molecular Formula C7H15NO2
Quantity 100 mg
Synonym L-2-Amino-4,4-dimethylpentanoic acid, (2S)-2-Amino-4,4-dimethylpentanoic acid
SMILES CC(C)(C[C@H](N)C(O)=O)C
Molecular Weight (g/mol) 145.2
Formula Weight 145.1103