Learn More
Keystone Cleanroom Products Gamma-Irradiated Heat-Seal Sterility Maintenance Bag
GREENER_CHOICE
Product Code. 17766879 Shop All Keystone Cleanroom Products Products
Click to view available options
Dimensions (L x W):
25.4 x 15.2 cm (6 x 10 in.)
30.5 x 20.3 cm (8 x 12 in.)
38.1 x 25.4 cm (10 x 15 in.)
50.8 x 30.5 cm (12 x 20 in.)
56 x 40.6 cm (16 x 22 in.)
76.2 x 40.6 cm (16 x 30 in.)
76.2 x 61 cm (24 x 30 in.)
94 x 63.5 cm (25 x 37 in.)
Unit Size:
Pack of 250
Pack of 500
Product Code. 17766879
Brand: Keystone Cleanroom Products 3220ST
Only null left
Add to basket
Add to basket
Bag used to cover wrapped items after sterilization to provide protection from environmental factors that could compromise sterility.
  • Made of a transparent, strong, 3 mil thick medical grade polyolefin material
  • Features a starter slit to facilitate easy opening of the cover without the need for cutting instruments which could damage the contents
  • Gamma irradiated
  • Lot controlled
  • Ready to use
  • Packaged in easy to open Tyvek™ peel pouches
  • Minimize contamination risks
  • Simplifies conformance to SOP's
  • Eliminate the need for wiping down contaminated writing instruments
Specifications
Autoclavable Not Autoclavable
Capacity (Metric) 6.51 L
Certifications/Compliance COC, Certificate of Irradiation
Closure Type Heat Seal
Color Clear
Disposable Single-use
Material Polyolefin
Sterility Sterile
Thickness (Metric) 76.2 μm
Latex Free Yes
Thickness (English) 3 mil
Dimensions (L x W) 50.8 x 30.5 cm (12 x 20 in.)
Length (English) 20 in.
Length (Metric) 50.8 cm
Width (English) 12 in.
Width (Metric) 30.5 cm
Product Type Cover