Learn More
Karl Hecht™ Assistent™ Test Tube Brushes
Description
- With cotton end
- Available for a range of different length and diameter tubes
Specifications
Specifications
| Product Type | Test Tube Brush |
| Diameter (Metric) Bristle | 26 mm |
| For Use With (Application) | For 25mm dia. tubes |
| Includes | Cotton end |
| Length (Metric) | 330 mm |
| Material | Galvanized Wire |
| Quantity | 1 Ea. |