missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GALNT3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15730411 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15730411 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. Consulta la politica di reso
Codice prodotto. 15730411 Fornitore Invitrogen™ N. del fornitore PA552458

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

Immunogen sequence: EESRMERNMK NKNKMLDLML EAVNNIKDAM PKMQIGAPVR QNIDAGERPC LQGYYTAAEL KPVLDRPPQD SNAPGASGKA FKTTNLSVEE QKEKERGEAK HC Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.

GALNT3 encodes UDP-GalNAc transferase 3, a member of the GalNAc-transferases family. This family transfers an N-acetyl galactosamine to the hydroxyl group of a serine or threonine residue in the first step of O-linked oligosaccharide biosynthesis. Individual GalNAc-transferases have distinct activities and initiation of O-glycosylation is regulated by a repertoire of GalNAc-transferases. The protein is highly homologous to other family members, however the enzymes have different substrate specificities.
TRUSTED_SUSTAINABILITY

Specifica

Antigen GALNT3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Galnt3
Gene Accession No. Q14435
Gene Alias GalNAc transferase 3; galNAc-T3; GALNT3; HFTC; HHS; polypeptide GalNAc transferase 3; polypeptide GalNAc-transferase T3; polypeptide N-acetylgalactosaminyltransferase 3; Polypeptide N-acetylgalactosaminyltransferase 3-like protein; pp-GaNTase 3; ppGaNTase-T3; protein-UDP acetylgalactosaminyltransferase 3; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 3; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3 (GalNAc-T3); UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3 (GalNAc-T3); polypeptide N-acetylgalactosaminyltransferase 3
Gene Symbols Galnt3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GALNT3. Recombinant protein control fragment (Product #RP-90125).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2591
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.