Learn More
GALNT3 293T Cell Overexpression Lysate (Denatured), Abnova
Description
Western Blotting
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | GALNT3 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | BC056246 |
| Molecular Weight (g/mol) | 15.9 |
| pH Range | 6.8 |
| Plasmid | pCMV-GALNT3 full-length |
| Quality Control Testing | Transient overexpression cell Lysate was tested with Anti-GALNT3 antibody (H00002591-B01) by western blotting |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Show More |
Safety and Handling
Product Identifier
- 100UL GALNT3 293T Cell Transient Overexpression Ly
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contain: Mercaptoethanol