missing translation for 'onlineSavingsMsg'
Learn More

Gallopamil, MedChemExpress

Product Code. p-200101335
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16476018

Brand: Medchem Express HY14276

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gallopamil (Methoxyverapamil), a methoxy derivative of Verapamil, is a phenylalkylamine calcium antagonist. Gallopamil inhibits acid secretion in a concentration-dependent manner with an IC50 of 10.9 μM. Gallopamil is a potent antiarrhythmic and vasodilator agent.

Gallopamil (Methoxyverapamil), a methoxy derivative of Verapamil, is a phenylalkylamine calcium antagonist. Gallopamil inhibits acid secretion in a concentration-dependent manner with an IC50 of 10.9 μM. Gallopamil is a potent antiarrhythmic and vasodilator agent.

Specifications

Chemical Name or Material Gallopamil
CAS 16662-47-8
Color White
Purity Grade Notes Research
Molecular Formula C28H40N2O5
Quantity 10 mg
Synonym Methoxyverapamil
Solubility Information DMSO : 100 mg/mL (206.34 mM; Need ultrasonic)
SMILES COC1=C(OC)C(OC)=CC(C(CCCN(C)CCC2=CC(OC)=C(OC)C=C2)(C#N)C(C)C)=C1
Molecular Weight (g/mol) 484.63
Formula Weight 484.63
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.