Learn More
Gallopamil, MedChemExpress
Product Code. 16476018
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16476018
Brand: Medchem Express HY14276
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Gallopamil (Methoxyverapamil), a methoxy derivative of Verapamil, is a phenylalkylamine calcium antagonist. Gallopamil inhibits acid secretion in a concentration-dependent manner with an IC50 of 10.9 μM. Gallopamil is a potent antiarrhythmic and vasodilator agent.
Gallopamil (Methoxyverapamil), a methoxy derivative of Verapamil, is a phenylalkylamine calcium antagonist. Gallopamil inhibits acid secretion in a concentration-dependent manner with an IC50 of 10.9 μM. Gallopamil is a potent antiarrhythmic and vasodilator agent.
Specifications
Chemical Name or Material Gallopamil
CAS 16662-47-8
Color White
Purity Grade Notes Research
Molecular Formula C28H40N2O5
Quantity 10 mg
Synonym Methoxyverapamil
Solubility Information DMSO : 100 mg/mL (206.34 mM; Need ultrasonic)
SMILES COC1=C(OC)C(OC)=CC(C(CCCN(C)CCC2=CC(OC)=C(OC)C=C2)(C#N)C(C)C)=C1
Molecular Weight (g/mol) 484.63
Formula Weight 484.63
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation