missing translation for 'onlineSavingsMsg'
Learn More

Galidesivir, MedChemExpress

Product Code. 16420038
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16420038

Brand: Medchem Express HY18649A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Galidesivir (BCX4430), an adenosine analog and a direct-acting antiviral agent, disrupts viral RNA-dependent RNA polymerase (RdRp) activity. Galidesivir is active in vitro against many RNA viral pathogens, including the filoviruses and emerging infectious agents such as MERS-CoV, SARS-CoV, and SARS-CoV-2. Galidesivir inhibits some negative-sense RNA viruses with EC50s ranging from ∼3 to ∼68 μM.

Galidesivir (BCX4430), an adenosine analog and a direct-acting antiviral agent, disrupts viral RNA-dependent RNA polymerase (RdRp) activity. Galidesivir is active in vitro against many RNA viral pathogens, including the filoviruses and emerging infectious agents such as MERS-CoV, SARS-CoV, and SARS-CoV-2. Galidesivir inhibits some negative-sense RNA viruses with EC50s ranging from ∼3 to ∼68 μM.

Specifications

Chemical Name or Material Galidesivir
Physical Form Solid
CAS 249503-25-1
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C11H15N5O3
For Use With (Application) COVID-19-anti-virus
Synonym BCX4430 Immucillin-A
Solubility Information H2O : 1.53 mg/mL (5.77 mM; Need ultrasonic and warming)
SMILES O[C@H]1[C@H](C2=CNC3=C2N=CN=C3N)N[C@H](CO)[C@H]1O
Molecular Weight (g/mol) 265.27
Formula Weight 265.27
Percent Purity 99.29%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.