missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-13 Mouse, Clone: PP13/1161, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 29124-MSM1-P0
This item is not returnable.
View return policy
Specifications
| Galectin-13 | |
| Cytoplasmic and Cell Surface | |
| PP13/1161 | |
| Unconjugated | |
| XP_016882204 | |
| Mouse | |
| 16kDa (monomer) ;32kDa (homodimer) | |
| Protein A/G Purified | |
| RUO | |
| 29124 | |
| IgG2b κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| LGALS13 | |
| Recombinant human Galectin-13 protein fragment (aa23-134) | |
| K-562 cells. Placenta or Spleen. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction