missing translation for 'onlineSavingsMsg'
Learn More

Gadoxetate Disodium, MedChemExpress

Product Code. 16419557
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16419557

Brand: Medchem Express HY16219

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gadoxetate Disodium (Gd-EOB-DTPA Disodium) is a contrast agent in magnetic resonance imaging (MRI) of the hepatobiliary system, which accumulates in normal, functioning hepatocytes. Gadoxetate Disodium (Gd-EOB-DTPA Disodium) is used to evaluate focal liver lesions, such as hepatocellular carcinoma or liver metastasis on T1-weighted imaging.

Gadoxetate Disodium (Gd-EOB-DTPA Disodium) is a contrast agent in magnetic resonance imaging (MRI) of the hepatobiliary system, which accumulates in normal, functioning hepatocytes. Gadoxetate Disodium (Gd-EOB-DTPA Disodium) is used to evaluate focal liver lesions, such as hepatocellular carcinoma or liver metastasis on T1-weighted imaging.

Specifications

Chemical Name or Material Gadoxetate Disodium
Physical Form Solid
CAS 135326-22-6
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H28GdN3Na2O11
For Use With (Application) Neuroscience-Neuromodulation
Synonym Gd-EOB-DTPA Disodium ZK 139834
Solubility Information DMSO : 24 mg/mL (33.07 mM; Need ultrasonic)
SMILES O=C1[O-][Gd+3]([N]2(C1)CC3=O)([O-]3)([O-]C4=O)([O-]C5=O)([N]6(C5)C7)([N](C4)(CC2)CC6CC8=CC=C(OCC)C=C8)[O-]C7=O.[Na+].[Na+]
Molecular Weight (g/mol) 725.71
Formula Weight 725.71
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.