missing translation for 'onlineSavingsMsg'
Learn More

G-5555, MedChemExpress

Product Code. 16430298
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16430298

Brand: Medchem Express HY196355mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress G-5555 is a potent p21-activated kinase 1 (PAK1) inhibitor with Kis of 3.7 nM and 11 nM for PAK1 and PAK2, respectively.

G-5555 is a potent p21-activated kinase 1 (PAK1) inhibitor with Kis of 3.7 nM and 11 nM for PAK1 and PAK2, respectively.

Specifications

Chemical Name or Material G-5555
CAS 1648863-90-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C25H25ClN6O3
Quantity 5 mg
Solubility Information DMSO : 25 mg/mL (50.71 mM; ultrasonic and warming and heat to 80°C)
SMILES ClC(C=C(C1=NC(C)=CC=C1)C=C2)=C2C3=CC4=CN=C(NC)N=C4N(C[C@@H]5OC[C@@H](N)CO5)C3=O
Molecular Weight (g/mol) 492.96
Formula Weight 492.96
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.