missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ FzM1.8

Product Code. 18746396
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18746396 10 mg 10mg
18798286 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18746396 Supplier Tocris Bioscience™ Supplier No. 6961/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Frizzled 4 allosteric agonist; exhibits biased siginaling; preserves stemness

FzM1.8 is a frizzled 4 allosteric agonist (pEC50 = 6.4). Potentiates the β-catenin pathway in the absence of any WNT ligand, by biasing FZD4 signaling toward the FZD4/PI3K axis. Preserves stemness and promotes proliferation of undifferentiated cells.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Hydroxy-5-[[(2-Naphthalenylamino)carbonyl]amino]benzoic acid
CAS 2204290-85-5
Quantity 10 mg
Molecular Formula C18H14N2O4
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.