Learn More
Carbolite™ Furnace with type 20/40 Temperature Sensor
Brand:  Carbolite™ HTF1808-230SN+TMP-3216P1BASE
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12973741
  • 33114.40 € / Each
Estimated Shipment: 01-07-2024
to see stock.
Add to basket
Specifications
Compliance to BSEN 61010
8 L
For use in quality control, heat treatment and research of ceramic materials.
830 mm
565 mm
650 mm
230 V 50/60 Hz 6200 w
6200 w
3216P1 Controller
Furnace
210 x 190 x 190 mm
190 mm
210 mm
190 mm
230 V
50/60 Hz