Learn More
Fumaric Acid Monomethyl Ester, TRC
Product Code. 30135495
Change view
Click to view available options
Quantity:
50 mg
10 g
25 g
Unit Size:
10g
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135495 25 g 25g
30138434 50 mg 50mg
30004194 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135495 Supplier Toronto Research Chemicals Supplier No. TRCF50039025G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2756-87-8
Molecular Formula C5 H6 O4
Molecular Weight (g/mol) 130.1
Synonym Monomethyl Fumarate, (2E)-2-Butenedioic acid 1-methyl ester, (E)-2-Butenedioic acid monomethyl ester, Fumaric acid methyl ester, Fumaric acid monomethyl ester, Methyl hydrogen fumarate, Monomethyl fumarate
IUPAC Name (E)-4-methoxy-4-oxobut-2-enoic acid
SMILES COC(=O)\C=C\C(=O)O
Specifications
Quantity 25 g
InChI Formula InChI=1S/C5H6O4/c1-9-5(8)3-2-4(6)7/h2-3H,1H3,(H,6,7)/b3-2+
Formula Weight 130.0266
Chemical Name or Material Monomethyl Fumarate