missing translation for 'onlineSavingsMsg'
Learn More

Fumarate hydratase-IN-1, MedChemExpress

Product Code. 16489985
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16489985

Brand: Medchem Express HY100004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fumarate hydratase-IN-1 (compound 2) is a cell-permeable fumarate hydratase inhibitor. Fumarate hydratase-IN-1 has antiproliferative activity against several cancer cell lines with a mean IC50 of 2.2 μM.

Fumarate hydratase-IN-1 (compound 2) is a cell-permeable fumarate hydratase inhibitor. Fumarate hydratase-IN-1 has antiproliferative activity against several cancer cell lines with a mean IC50 of 2.2 μM.

Specifications

Chemical Name or Material Fumarate hydratase-IN-1
CAS 1644060-37-6
Color White
Purity Grade Notes Research
Molecular Formula C27H30N2O4
Quantity 2 mg
Solubility Information DMSO : 50 mg/mL (111.97 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(OCC)[C@]1([C@H](CC(NC)=O)C2=O)C(N2CC(C=C3)=CC=C3C4=CC=CC=C4)=CCCC1
Molecular Weight (g/mol) 446.54
Formula Weight 446.54
Percent Purity 99.63%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.