Learn More
Ekastu Safety™ CLARO Full-View Goggles
Product Code. 10185930
Change view
Click to view available options
Lens Tint:
Clear
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint Unit Size
10185930 Clear Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10185930 Supplier Ekastu Safety™ Supplier No. 277381
  • ONLINE EXCLUSIVE PRICE
    8.65 €
    7.90 € / Pack of 1
    Save 0.75 €
    8% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Manufactured from soft plastic to protect against dust and liquid splashes. EKASTU Safety™ CLARO Full-View Goggles have a clear, scratch-proof, exchangeable anti-fog lens with 99.99% UV protection and innovative UV 380 technology.
  • DIN EN 166 1 3 4 9 – BT
  • Full view goggles made of soft plastic against dust and liquid splashes
  • Very high mechanical stability
  • Clear, scratch-proof, exchangeable non-fog lens
  • Wide supporting surface
  • Large field of vision
  • Maximum wearing comfort
  • 99.99% UV protection with innovative UV 380 technology
  • Also suitable for people who wear glasses and respiratory masks from EKASTU
Specifications
Certifications/Compliance DIN EN 166 1 3 4 9 – BT
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Headband Material Textile Elastic
No. per Pack 1 Ea.
Material Polycarbonate lens
Frame Material PVC
UV Protection Yes