missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Fucosyl GM1 Antibody (BMS-986012), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28503-100ug
This item is not returnable.
View return policy
Description
Fucosyl GM1 Monoclonal antibody specifically detects Fucosyl GM1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Fucosyl GM1 | |
| Monoclonal | |
| Unconjugated | |
| Q10981 | |
| Fucosyl GM1 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| BMS-986012 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Human | |
| Protein A purified | |
| RUO | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction