Learn More
Novus Biologicals™ Fucosyl GM1 Antibody (BMS-986012), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28503-50ug
 View more versions of this product
Product Code. 30045223
  • 213.00 € / 50µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Fucosyl GM1 Monoclonal antibody specifically detects Fucosyl GM1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fucosyl GM1
Monoclonal
Unconjugated
Q10981
Fucosyl GM1
50 μg
Primary
Human
Purified
Flow Cytometry, ELISA, Functional Assay
BMS-986012
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Human
Protein A purified
RUO
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1