missing translation for 'onlineSavingsMsg'
Learn More

FTSJ1 Antibody, DyLight 488, Novus Biologicals™

Product Code. 30027550 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30027550 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30027550 Supplier Novus Biologicals Supplier No. NBP326088G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FTSJ1 Polyclonal antibody specifically detects FTSJ1 in Human samples. It is validated for Western Blot, Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen FTSJ1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias CDLIV, cell division protein, EC 2.1.1.-, FtsJ homolog 1 (E. coli), JM23, mental retardation, X-linked 44, mental retardation, X-linked 9, MRX44, Protein ftsJ homolog 1, putative ribosomal RNA methyltransferase 1, rRNA (uridine-2'-O-)-methyltransferase, SPB1MRX9, TRM7
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human FTSJ1
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24140
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.