missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTO Antibody (4G9), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00 € - 533.00 €
Specifications
| Antigen | FTO |
|---|---|
| Clone | 4G9 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043274
|
Novus Biologicals
NBP3-26565-50ul |
50 μL |
343.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043667
|
Novus Biologicals
NBP3-26565-100ul |
100 μL |
533.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FTO Monoclonal antibody specifically detects FTO in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
| FTO | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149 | |
| A synthesized peptide derived from Human FTO [UniProt Q9C0B1] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 4G9 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism, Neuroscience | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 79068 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title