missing translation for 'onlineSavingsMsg'
Learn More

FT113, MedChemExpress

Product Code. 16417217
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16417217

Brand: Medchem Express HY111551

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FT113 is a potent and orally active fatty acid synthase (FASN) inhibitor, with an IC50 of 213 nM for full-length recombinant human FASN enzyme. In cell-based assay, FT113 blocks FASN activity in BT474 cells (IC50, 90 nM). FT113 shows anti-proliferative activity, and exhibits anti-cancer activity both in vitro and in vivo.

FT113 is a potent and orally active fatty acid synthase (FASN) inhibitor, with an IC50 of 213 nM for full-length recombinant human FASN enzyme. In cell-based assay, FT113 blocks FASN activity in BT474 cells (IC50, 90 nM). FT113 shows anti-proliferative activity, and exhibits anti-cancer activity both in vitro and in vivo.

Specifications

Chemical Name or Material FT113
CAS 1630808-89-7
Color White
Purity Grade Notes Research
Molecular Formula C22H20FN3O4
Quantity 5 mg
Solubility Information DMSO : 62.5 mg/mL (152.66 mM; Need ultrasonic)
SMILES O=C(C1(CC1)O)N2CCN(C(C3=CC=C(C4=NC5=C(C=C(F)C=C5)O4)C=C3)=O)CC2
Molecular Weight (g/mol) 409.41
Formula Weight 409.41
Percent Purity 99.61%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.