missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FRK Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (96%), Rat (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FRK (PTK5) is a nuclear protein and may function during G1 and S phase of the cell cycle, suppressing growth. The human FRK gene maps to chromosome 6q21-q22.3 and encodes a 505 amino acid protein. The protein belongs to the TYR family of protein kinases. This tyrosine kinase is a nuclear protein and may function during G1 and S phase of the cell cycle and suppress growth.
Specifications
Specifications
| Antigen | FRK |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.98 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | FRK |
| Gene Accession No. | P42685 |
| Gene Alias | B-cell src-homology tyrosine kinase; beta-cell Src-homology tyrosine kinase; BSK; BSK/IYK; C85044; EC 2.7.1.112; EC 2.7.10.2; FRK; fyn related Src family tyrosine kinase; fyn-related kinase; fyn-related Src family tyrosine kinase; GASK; Gastrointestinal-associated kinase; gastrointestinal-associated tyrosine kinase; GTK; intestine tyrosine kinase; Iyk; Nuclear tyrosine protein kinase RAK; protein-tyrosine kinase 5; PTK5; PTK5 protein tyrosine kinase 5; RAK; src related tyrosine kinase; tyrosine-protein kinase FRK |
| Gene Symbols | FRK |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?