missing translation for 'onlineSavingsMsg'
Learn More

Frizzled-3 Antibody (169310) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 18550216 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18550216 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18550216 Supplier Novus Biologicals Supplier No. FAB1001I

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rat Monoclonal Antibody

Frizzled-3 Monoclonal antibody specifically detects Frizzled-3 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Frizzled-3
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 169310
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias frizzled (Drosophila) homolog 3, frizzled homolog 3 (Drosophila), frizzled-3, Fz-3, hFz3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-3, Met1-Val157, Accession # Q61086
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7976
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.