Learn More
Eppendorf™ Freezer Rack: Innova™ U360, U535, U725/-G, CryoCube™ F740 series (3-comp., Std),
Description
Freezer Rack: Innova™ U360, U535, U725, U725-G, CryoCube™ F740 series (3-compartment, Standard), 4 in/102 mm, drawer, stainless steel- Rack for 102 mm/4 in boxes- Max. box size [mm]: 136 x 136- Drawer rack- Ext. dimensions of rack (DxWxH) [mm]: 580 x 140 x 324 (22.8 x 5.5 x 12.8 in)- Max. box number per rack: 12 (4 x 3)- Stainless steel- For ULT freezer: Innova U360 (max. 9 racks), Innova U535 (max. 12 racks), Innova U725 and U725-G (max. 18 racks), CryoCube F740 series (3-compartment version; max. 18 racks)- For ULT freezer: Innova U360 (max. 108 boxes), Innova U535 (max. 144 boxes), Innova U725 and 725-G (max. 216 boxes), CryoCube F740 series (3-compartment version; max. 216 boxes)
Specifications
Specifications
| Product Type | Freezer Rack |