Learn More
Eppendorf™ Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, lock, 4 in/102 mm
Description
Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, locking rod, 4 in/102 mm, side access, stainless steel- Rack for 102 mm/4.0 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 148 x 141 x 741 (5.8 x 5.6 x 29.2 in)- Max. box number per rack: 7 (7 x 1)\n- Stainless steel- For ULT freezer: Innova C580 (max. 32 racks), Innova C760 (max. 40 racks), CryoCube™ FC660 series (max. 36 racks)- For ULT freezer: Innova C580 (max. 224 boxes), Innova C760 (max. 200 boxes), CryoCube™ FC660 series (max. 252 boxes)- With locking rod for increased sample safety
Specifications
Specifications
| Product Type | Freezer Rack |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.