Learn More
Eppendorf™ Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, lock, 4 in/102 mm
Product Code. 15763225
This item is not returnable. View return policy
Product Code. 15763225
Brand: Eppendorf™ 6001000411
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, locking rod, 4 in/102 mm, side access, stainless steel- Rack for 102 mm/4.0 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 148 x 141 x 741 (5.8 x 5.6 x 29.2 in)- Max. box number per rack: 7 (7 x 1)\n- Stainless steel- For ULT freezer: Innova C580 (max. 32 racks), Innova C760 (max. 40 racks), CryoCube™ FC660 series (max. 36 racks)- For ULT freezer: Innova C580 (max. 224 boxes), Innova C760 (max. 200 boxes), CryoCube™ FC660 series (max. 252 boxes)- With locking rod for increased sample safety
TRUSTED_SUSTAINABILITY
Specifications
Product Type Freezer Rack