missing translation for 'onlineSavingsMsg'
Learn More

FRAX597, MedChemExpress

Product Code. 16432707
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16432707

Brand: Medchem Express HY15542A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FRAX597 is a potent group I p21-activated Kinases (PAKs) inhibitor with IC50 of 8, 13 and 19 nM for PAK1, 2 and 3.

FRAX597 is a potent group I p21-activated Kinases (PAKs) inhibitor with IC50 of 8, 13 and 19 nM for PAK1, 2 and 3.

Specifications

Chemical Name or Material FRAX597
Physical Form Solid
CAS 1286739-19-2
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C29H28ClN7OS
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 14.29 mg/mL (25.60 mM; Need ultrasonic)
SMILES O=C1C(C2=CC=C(C3=CN=CS3)C=C2Cl)=CC4=CN=C(NC5=CC=C(N6CCN(C)CC6)C=C5)N=C4N1CC
Molecular Weight (g/mol) 558.1
Formula Weight 558.1
Percent Purity 98.55%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.