missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FRAT1 Monoclonal antibody specifically detects FRAT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
Specifications
| Antigen | FRAT1 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Clone | SR1483 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Formulation | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gene Alias | FLJ97193, FRAT-1, frequently rearranged in advanced T-cell lymphomas, Frequently rearranged in advanced T-cell lymphomas 1, proto-oncogene FRAT1 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human FRAT1 (Uniprot #: Q92837) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?