missing translation for 'onlineSavingsMsg'
Learn More

FRAT1 Antibody (SR1483), Novus Biologicals™

Product Code. 18667594 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18667594 50 μg 50µL
18696784 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18667594 Supplier Novus Biologicals Supplier No. NBP32254350ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FRAT1 Monoclonal antibody specifically detects FRAT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen FRAT1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone SR1483
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias FLJ97193, FRAT-1, frequently rearranged in advanced T-cell lymphomas, Frequently rearranged in advanced T-cell lymphomas 1, proto-oncogene FRAT1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FRAT1 (Uniprot #: Q92837)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 10023
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.