missing translation for 'onlineSavingsMsg'
Learn More

FR 180204, MedChemExpress

Product Code. 16437777
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16437777

Brand: Medchem Express HY12275

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FR 180204 is an ATP-competitive and selective ERK inhibitor. FR 180204 inhibits ERK1 and ERK2 with IC50s of 0.51 μM (Ki=0.31 μM) and 0.33 μM (Ki=0.14 μM), respectively.

FR 180204 is an ATP-competitive and selective ERK inhibitor. FR 180204 inhibits ERK1 and ERK2 with IC50s of 0.51 μM (Ki=0.31 μM) and 0.33 μM (Ki=0.14 μM), respectively.

Specifications

Chemical Name or Material FR 180204
Physical Form Solid
CAS 865362-74-9
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C18H13N7
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 50 mg/mL (152.75 mM)
SMILES NC1=NNC2=NN=C(C3=C4C=CC=CN4N=C3C5=CC=CC=C5)C=C21
Molecular Weight (g/mol) 327.34
Formula Weight 327.34
Percent Purity 99.47%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.