missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FPGS Recombinant Rabbit Monoclonal Antibody (30C11)

Product Code. 17910543
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17910543 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17910543 Supplier Invitrogen™ Supplier No. MA550184
  • ONLINE EXCLUSIVE PRICE
    446.00 €
    379.10 € / 100µL
    Save 66.90 €
    15% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

FPGS Recombinant Monoclonal Antibody for Flow, ELISA

Folylpoly-gamma-glutamate synthetase (FPGS) catalyzes the addition of several equivalents of glutamic acid to the gamma-carboxyl group in the side chain of folate cofactors and analogs. Folylpoly-gamma-glutamate synthetase has three functions in folate homeostasis in mammals: polyglutamation prevents efflux of folate cofactors from the cell, it increases the binding of folate cofactors to some of the enzymes of folate interconversion and biosynthesis, and it appears to allow the accumulation of folates in the mitochondria that are required for glycine synthesis. Folylpoly-gamma-glutamate synthetase is widely expressed in human tumors and is tightly linked either to proliferation or to a lack of differentiation. The cytotoxicity of both thymidylate synthase and purine inhibitors requires continued inhibition of target for greater than one generation time, so that the integrative function of FPGS adds considerably to the efficiency of folate antimetabolites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FPGS
Applications ELISA, Flow Cytometry
Classification Recombinant Monoclonal
Clone 30C11
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FPGS
Gene Accession No. Q05932
Gene Alias AA408187; folylpoly-gamma-glutamate synthetase; folylpolyglutamate synthase; folylpolyglutamate synthase, mitochondrial; folylpolyglutamyl synthetase; FPGS; Tetrahydrofolate synthase; tetrahydrofolylpolyglutamate synthase
Gene Symbols FPGS
Host Species Rabbit
Immunogen A synthesized peptide derived from Human FPGS.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2356
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.